siRNA Details
| Gene Symbol: FOXF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001451.2 | NP_001442.2 | 1 | 189 | ||
| GenBank mRNA | AK314167.1 | - | 1 | 196 | ||
| BC089442.1 | - | 1 | 178 | 30558276 | ||
| DB011539.1 | - | 1 | 196 | |||
| U13219.1 | - | 1 | 164 | |||

FOXF1
Genomic Map