siRNA Details
Gene Symbol: FOXE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012186.2 | NP_036318.1 | 1 | 1331 | ||
GenBank mRNA | AF275722.1 | - | 1 | 1342 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FOXE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012186.2 | NP_036318.1 | 1 | 1331 | ||
GenBank mRNA | AF275722.1 | - | 1 | 1342 |