siRNA Details
| Gene Symbol: FOXE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012186.2 | NP_036318.1 | 1 | 1668 | ||
| GenBank mRNA | AF275722.1 | - | 1 | 1679 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOXE3| Gene Symbol: FOXE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012186.2 | NP_036318.1 | 1 | 1668 | ||
| GenBank mRNA | AF275722.1 | - | 1 | 1679 | ||