siRNA Details
| Gene Symbol: FUT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000149.3 | NP_000140.1 | 3 | 977 | ||
| NM_001097639.1 | NP_001091108.1 | 3 | 629 | |||
| NM_001097640.1 | NP_001091109.1 | 3 | 575 | |||
| NM_001097641.1 | NP_001091110.1 | 2 | 436 | |||
| XM_011527865.1 | XP_011526167.1 | 3 | 955 | |||
| XM_011527866.1 | XP_011526168.1 | 3 | 616 | |||
| XM_011527867.1 | XP_011526169.1 | 3 | 585 | |||
| GenBank mRNA | AF131913.1 | - | 2 | 508 | ||
| AK291852.1 | - | 3 | 604 | |||
| AK313575.1 | - | 3 | 626 | |||
| BC074836.2 | - | 2 | 381 | 30915328 | ||
| BC074837.2 | - | 2 | 381 | 30915548 | ||
| BC108675.1 | - | 2 | 433 | 5187745 | ||
| DA876997.1 | - | 3 | 550 | |||
| U27326.1 | - | 3 | 568 | |||
| U27327.1 | - | 3 | 622 | |||
| U27328.1 | - | 4 | 971 | |||
| X53578.1 | - | 2 | 411 | |||

FUT3
Genomic Map