siRNA Details
| Gene Symbol: FUT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002034.2 | NP_002025.2 | 2 | 233 | ||
| GenBank mRNA | BC140905.1 | - | 2 | 229 | 9056717 | |
| U27329.1 | - | 2 | 233 | |||
| U27330.1 | - | 2 | 233 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FUT5| Gene Symbol: FUT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002034.2 | NP_002025.2 | 2 | 233 | ||
| GenBank mRNA | BC140905.1 | - | 2 | 229 | 9056717 | |
| U27329.1 | - | 2 | 233 | |||
| U27330.1 | - | 2 | 233 | |||