siRNA Details
| Gene Symbol: GAS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286205.1 | NP_001273134.1 | 5 | 764 | ||
| NM_001286209.1 | NP_001273138.1 | 5 | 656 | |||
| NM_001481.2 | NP_001472.1 | 5 | 641 | |||
| XM_005256304.4 | XP_005256361.1 | 6 | 1296 | |||
| XM_005256309.4 | XP_005256366.1 | 7 | 1021 | |||
| XM_006721175.3 | XP_006721238.1 | 5 | 694 | |||
| XM_011522990.2 | XP_011521292.1 | 5 | 715 | |||
| XM_011522991.1 | XP_011521293.1 | 6 | 870 | |||
| XM_011522992.2 | XP_011521294.1 | 4 | 663 | |||
| XM_017023122.1 | XP_016878611.1 | 7 | 1021 | |||
| XM_017023123.1 | XP_016878612.1 | 5 | 720 | |||
| GenBank mRNA | AF050079.1 | - | 5 | 640 | ||
| AK293958.1 | - | 5 | 671 | |||
| AK294050.1 | - | 4 | 490 | |||
| AK297857.1 | - | 5 | 624 | |||
| AK303505.1 | - | 5 | 697 | |||
| AK315261.1 | - | 5 | 589 | |||
| BC036781.1 | - | 7 | 2445 | |||
| BC104785.1 | - | 5 | 626 | 8143788 | ||
| BC112121.1 | - | 5 | 626 | 8327589 | ||
| CR749499.1 | - | 4 | 674 | |||

GAS8
Genomic Map