siRNA Details
| Gene Symbol: GH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002059.4 | NP_002050.1 | 4 | 513 | ||
| NM_022556.3 | NP_072050.1 | 4 | 468 | |||
| NM_022557.3 | NP_072051.1 | 4 | 513 | |||
| NM_022558.3 | NP_072052.1 | 4 | 513 | |||
| GenBank mRNA | AB451332.1 | - | 3 | 327 | ||
| AB451477.1 | - | 3 | 327 | |||
| AF006060.1 | - | 4 | 339 | |||
| AF006061.1 | - | 3 | 372 | |||
| AK312194.1 | - | 4 | 435 | |||
| AU139016.1 | - | 4 | 513 | |||
| BC020760.1 | - | 4 | 434 | 4720475 | ||
| CR541902.1 | - | 3 | 372 | |||
| CR541924.1 | - | 3 | 372 | |||
| J03756.1 | - | 4 | 433 | |||
| M38451.1 | - | 4 | 384 | |||

GH2
Genomic Map