siRNA Details
| Gene Symbol: GIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004123.2 | NP_004114.1 | 3 | 206 | ||
| GenBank mRNA | BC069100.1 | - | 3 | 206 | 7216914 | |
| BC069663.1 | - | 3 | 206 | 7262542 | ||
| BC069686.1 | - | 3 | 206 | 7262554 | ||
| BC069746.1 | - | 3 | 206 | 7262566 | ||
| BC096146.1 | - | 3 | 205 | 40002916 | ||
| BC096147.1 | - | 3 | 205 | 40002922 | ||
| BC096148.3 | - | 3 | 205 | 40002923 | ||
| BC096149.3 | - | 3 | 205 | 40002925 | ||
| E02006.1 | - | 3 | 208 | |||
| M18185.1 | - | 3 | 206 | |||

GIP
Genomic Map