siRNA Details
| Gene Symbol: GYPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304382.1 | NP_001291311.1 | 6 | 464 | ||
| NM_002100.5 | NP_002091.3 | 5 | 384 | |||
| XM_011531905.2 | XP_011530207.1 | 6 | 671 | |||
| XM_017008137.1 | XP_016863626.1 | 5 | 2718 | |||
| GenBank mRNA | BC069310.1 | - | 5 | 376 | 7262104 | |
| BC121077.1 | - | 5 | 360 | 40119713 | ||
| BC121078.1 | - | 5 | 360 | 40119716 | ||
| EU338223.1 | - | 4 | 264 | |||
| EU338224.1 | - | 6 | 487 | |||
| EU338225.1 | - | 6 | 456 | |||
| EU338226.1 | - | 6 | 421 | |||
| EU338227.1 | - | 3 | 169 | |||
| EU338228.1 | - | 4 | 264 | |||
| EU338229.1 | - | 6 | 487 | |||
| EU338230.1 | - | 5 | 392 | |||
| EU338235.1 | - | 4 | 264 | |||
| EU338236.1 | - | 6 | 487 | |||
| EU338237.1 | - | 7 | 580 | |||
| EU338238.1 | - | 3 | 225 | |||
| U05255.1 | - | 4 | 231 | |||
| X08055.1 | - | 7 | 462 | |||

GYPB
Genomic Map