siRNA Details
| Gene Symbol: HIST1H1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005322.2 | NP_005313.1 | 1 | 248 | ||
| GenBank mRNA | BC069101.1 | - | 1 | 248 | 7216915 | |
| BC101581.1 | - | 1 | 221 | 8069087 | ||
| BC101583.1 | - | 1 | 221 | 8069089 | ||

HIST1H1B
Genomic Map