siRNA Details
| Gene Symbol: HBBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001589.1 | 3 | 433 | |||
| GenBank mRNA | N55205.1 | - | 1 | 225 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HBBP1| Gene Symbol: HBBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001589.1 | 3 | 433 | |||
| GenBank mRNA | N55205.1 | - | 1 | 225 | ||