siRNA Details
| Gene Symbol: RUFY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103475.1 | 19 | 2259 | |||
| NR_103476.1 | 19 | 2084 | ||||
| GenBank mRNA | AB040970.1 | - | 19 | 2020 | ||
| AF461266.1 | - | 18 | 1990 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: RUFY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103475.1 | 19 | 2259 | |||
| NR_103476.1 | 19 | 2084 | ||||
| GenBank mRNA | AB040970.1 | - | 19 | 2020 | ||
| AF461266.1 | - | 18 | 1990 | |||