siRNA Details
| Gene Symbol: HSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001537.3 | NP_001528.1 | 4 | 1908 | ||
| GenBank mRNA | AK026575.1 | - | 1 | 1106 | ||
| AK172732.1 | - | 4 | 1773 | |||
| BX537440.1 | - | 4 | 1797 | |||
| BX648972.1 | - | 4 | 1791 | |||
| DB519225.1 | - | 1 | 396 | |||

HSBP1
Genomic Map