siRNA Details
| Gene Symbol: HSPE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002157.2 | NP_002148.1 | 2 | 583 | ||
| GenBank mRNA | AK312104.1 | - | 2 | 179 | ||
| BC023518.2 | - | 2 | 257 | 4304224 | ||
| BG188453.1 | - | 1 | 144 | |||
| CN264788.1 | - | 2 | 588 | |||
| CR407688.1 | - | 1 | 143 | |||
| U07550.1 | - | 2 | 184 | |||
| X75821.1 | - | 1 | 143 | |||
| Gene Symbol: HSPE1-MOB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001202485.1 | NP_001189414.1 | 2 | 583 | ||
| GenBank mRNA | BJ992310.1 | - | 2 | 201 | ||
| CR990871.1 | - | 2 | 167 | |||

HSPE1
Genomic Map