siRNA Details
| Gene Symbol: HTR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000871.2 | NP_000862.1 | 3 | 1717 | ||
| GenBank mRNA | AB209129.1 | - | 1 | 6369 | ||
| AY429105.1 | - | 3 | 1250 | |||
| BC074995.2 | - | 3 | 1284 | 30915396 | ||
| BC074996.2 | - | 3 | 1284 | 30915608 | ||
| L41147.1 | - | 3 | 1717 | |||
More Information
Set Membership: |
|

HTR6
Genomic Map