siRNA Details
| Gene Symbol: IDH3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004135.3 | NP_004126.1 | 11 | 1139 | ||
| NM_174869.2 | NP_777358.1 | 11 | 1139 | |||
| GenBank mRNA | AB209206.1 | - | 5 | 3616 | ||
| BC000933.2 | - | 11 | 985 | 3447003 | ||
| BC001902.1 | - | 11 | 980 | 3543839 | ||
| U40272.1 | - | 11 | 1134 | |||
| U69268.1 | - | 7 | 690 | |||
| U69269.1 | - | 4 | 358 | |||
| Z68907.1 | - | 11 | 1021 | |||

IDH3G
Genomic Map