siRNA Details
| Gene Symbol: IGFBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000597.2 | NP_000588.2 | 2, 3 | 807 | ||
| NM_001313990.1 | NP_001300919.1 | 2, 3 | 365 | |||
| NM_001313992.1 | NP_001300921.1 | 2, 3 | 684 | |||
| NM_001313993.1 | NP_001300922.1 | 2, 3 | 423 | |||
| GenBank mRNA | A09809.1 | - | 2, 3 | 806 | ||
| AL559513.3 | - | 2, 3 | 814 | |||
| BC004312.1 | - | 2, 3 | 781 | 3627826 | ||
| BC009902.1 | - | 2, 3 | 783 | 3509749 | ||
| BC012769.1 | - | 2, 3 | 781 | 3689990 | ||
| BC071967.1 | - | 2, 3 | 794 | 6650239 | ||
| BE735562.1 | - | 2, 3 | 365 | |||
| CN431340.1 | - | 2, 3 | 423 | |||
| M35410.1 | - | 2, 3 | 750 | |||
| X16302.1 | - | 2, 3 | 804 | |||

IGFBP2
Genomic Map