siRNA Details
| Gene Symbol: FOXK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004514.3 | NP_004505.2 | 4 | 988 | ||
| GenBank mRNA | U58196.1 | - | 4 | 999 | ||
| U58197.1 | - | 4 | 999 | |||
| U58198.1 | - | 4 | 999 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOXK2| Gene Symbol: FOXK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004514.3 | NP_004505.2 | 4 | 988 | ||
| GenBank mRNA | U58196.1 | - | 4 | 999 | ||
| U58197.1 | - | 4 | 999 | |||
| U58198.1 | - | 4 | 999 | |||