siRNA Details
| Gene Symbol: ISLR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005545.3 | NP_005536.1 | 2 | 1208 | ||
| NM_201526.1 | NP_958934.1 | 2 | 1024 | |||
| GenBank mRNA | AB003184.1 | - | 2 | 1024 | ||
| AB024536.1 | - | 2 | 1208 | |||
| AK000817.1 | - | 2 | 1209 | |||
| AK074668.1 | - | 2 | 1616 | |||
| AK223267.1 | - | 2 | 1040 | |||
| AK225606.1 | - | 3 | 660 | |||
| AK297730.1 | - | 2 | 788 | |||
| AY358871.1 | - | 2 | 1284 | |||
| BC022478.1 | - | 2 | 1013 | 4794738 | ||
| BC111013.1 | - | 2 | 1118 | 5763157 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001195431, NM_012043, AB024538, BC006602.

ISLR
Genomic Map