siRNA Details
| Gene Symbol: ISLR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005545.3 | NP_005536.1 | 2 | 1431 | ||
| NM_201526.1 | NP_958934.1 | 2 | 1247 | |||
| GenBank mRNA | AB003184.1 | - | 2 | 1247 | ||
| AB024536.1 | - | 2 | 1431 | |||
| AK000817.1 | - | 2 | 1432 | |||
| AK074668.1 | - | 2 | 1839 | |||
| AK223267.1 | - | 2 | 1263 | |||
| AK225606.1 | - | 3 | 883 | |||
| AK297730.1 | - | 2 | 1011 | |||
| AY358871.1 | - | 2 | 1507 | |||
| BC022478.1 | - | 2 | 1236 | 4794738 | ||
| BC111013.1 | - | 2 | 1341 | 5763157 | ||

ISLR
Genomic Map