siRNA Details
| Gene Symbol: KRT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005556.3 | NP_005547.3 | 8 | 1353 | ||
| XM_017019294.1 | XP_016874783.1 | 8 | 1079 | |||
| GenBank mRNA | AF509887.1 | - | 8 | 1282 | ||
| AJ238246.1 | - | 1741 | ||||
| AK025881.1 | - | 8 | 1280 | |||
| AK128505.1 | - | 7 | 4021 | |||
| AK128848.1 | - | 9 | 1208 | |||
| AK129924.1 | - | 9 | 1287 | |||
| AK225498.1 | - | 8 | 1276 | |||
| BC002700.2 | - | 8 | 1268 | 3610347 | ||
| BC009521.1 | - | 4 | 440 | |||
| BE740515.1 | - | 4 | 484 | |||
| X03212.1 | - | 8 | 1287 | |||

KRT7
Genomic Map