siRNA Details
| Gene Symbol: KRT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005556.3 | NP_005547.3 | 4, 5 | 811 | ||
| XM_011538325.1 | XP_011536627.1 | 4, 5 | 764 | |||
| XM_017019294.1 | XP_016874783.1 | 4, 5 | 537 | |||
| GenBank mRNA | AF509887.1 | - | 4, 5 | 740 | ||
| AJ238246.1 | - | 4, 5 | 745 | |||
| AK025881.1 | - | 4, 5 | 738 | |||
| AK128505.1 | - | 4, 5 | 630 | |||
| AK128848.1 | - | 5, 6 | 666 | |||
| AK129924.1 | - | 5, 6 | 745 | |||
| AK225498.1 | - | 4, 5 | 734 | |||
| BC002700.2 | - | 4, 5 | 726 | 3610347 | ||
| BC107082.1 | - | 1, 2 | 70 | |||
| BE744321.1 | - | 388 | ||||
| X03212.1 | - | 4, 5 | 745 | |||
| Z36852.1 | - | 2, 3 | 151 | |||

KRT7
Genomic Map