siRNA Details
| Gene Symbol: SMCP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030663.2 | NP_109588.2 | 2 | 400 | ||
| GenBank mRNA | AK312020.1 | - | 2 | 400 | ||
| BC014593.2 | - | 2 | 400 | 4824172 | ||
| BC016744.2 | - | 2 | 383 | 4829283 | ||
| X89960.1 | - | 2 | 380 | |||

SMCP
Genomic Map