siRNA Details
| Gene Symbol: MIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002415.1 | NP_002406.1 | 3 | 412 | ||
| GenBank mRNA | AF469046.1 | - | 3 | 315 | ||
| AK311929.1 | - | 3 | 407 | |||
| BC000447.2 | - | 3 | 396 | 2821346 | ||
| BC007676.1 | - | 3 | 380 | 3605756 | ||
| BC008914.2 | - | 3 | 366 | 3615896 | ||
| BC013976.2 | - | 3 | 411 | 4130651 | ||
| BC022414.1 | - | 3 | 413 | 4243461 | ||
| BC053376.1 | - | 3 | 349 | 6047427 | ||
| BT007148.1 | - | 3 | 315 | |||
| CR407644.1 | - | 3 | 315 | |||
| CR456520.1 | - | 3 | 335 | |||
| CR541651.1 | - | 3 | 315 | |||
| JQ846015.1 | - | 3 | 315 | |||
| L10612.1 | - | 3 | 389 | |||
| M95775.1 | - | 3 | 289 | |||
| Z23063.1 | - | 3 | 315 | |||
| Gene Symbol: MIF-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038911.1 | 3 | 825 | |||
| GenBank mRNA | BC036909.1 | - | 3 | 825 | ||

MIF
Genomic Map