siRNA Details
| Gene Symbol: CEACAM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002483.6 | NP_002474.4 | 6 | 2076 | ||
| XM_011526990.1 | XP_011525292.1 | 5 | 2058 | |||
| GenBank mRNA | BC005008.1 | - | 6 | 1997 | 3640231 | |
| M18216.1 | - | 6 | 2001 | |||
| M18728.1 | - | 6 | 2007 | |||

CEACAM6
Genomic Map