siRNA Details
| Gene Symbol: CEACAM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002483.6 | NP_002474.4 | 3 | 723 | ||
| XM_011526990.1 | XP_011525292.1 | 3 | 822 | |||
| GenBank mRNA | AK312542.1 | - | 3 | 708 | ||
| BC005008.1 | - | 3 | 644 | 3640231 | ||
| BT009774.1 | - | 3 | 604 | |||
| M18216.1 | - | 3 | 650 | |||
| M18728.1 | - | 3 | 654 | |||
| M29541.1 | - | 3 | 697 | |||

CEACAM6
Genomic Map