siRNA Details
| Gene Symbol: NDUFB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004546.2 | NP_004537.1 | 2, 3 | 301 | ||
| GenBank mRNA | AF050639.1 | - | 2, 3 | 294 | ||
| AF067166.1 | - | 2, 3 | 301 | |||
| BC001168.1 | - | 2, 3 | 268 | |||
| BC063026.1 | - | 3, 4 | 466 | 6064434 | ||
| CR542121.1 | - | 2, 3 | 237 | |||

NDUFB2
Genomic Map