siRNA Details
| Gene Symbol: NDUFB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199987.1 | NP_001186916.1 | 3 | 395 | ||
| NM_002493.4 | NP_002484.1 | 4 | 488 | |||
| NM_182739.2 | NP_877416.1 | 3 | 443 | |||
| GenBank mRNA | AF035840.1 | - | 4 | 432 | ||
| AF067167.1 | - | 4 | 461 | |||
| AK092649.1 | - | 1 | 1880 | |||
| AK289702.1 | - | 4 | 442 | |||
| AW160372.1 | - | 3 | 326 | |||
| BC009801.1 | - | 4 | 467 | 4334414 | ||
| BC016708.1 | - | 4 | 502 | |||
| BC029247.1 | - | 4 | 433 | 5171627 | ||
| BG220924.1 | - | 2 | 100 | |||
| BI669356.1 | - | 3 | 422 | |||
| BP208846.1 | - | 4 | 486 | |||
| BU952486.1 | - | 1 | 371 | |||
| CR456920.1 | - | 4 | 364 | |||

NDUFB6
Genomic Map