siRNA Details
| Gene Symbol: NDUFB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284367.1 | NP_001271296.1 | 5 | 827 | ||
| NM_001284368.1 | NP_001271297.1 | 5 | 526 | |||
| NM_005004.3 | NP_004995.1 | 5 | 617 | |||
| GenBank mRNA | AA782554.1 | - | 1 | 138 | ||
| AF044958.1 | - | 5 | 567 | |||
| AF077028.1 | - | 5 | 564 | |||
| AF115968.1 | - | 5 | 573 | |||
| AI282606.1 | - | 1 | 111 | |||
| AK289579.1 | - | 5 | 558 | |||
| BC000466.2 | - | 5 | 558 | 2822781 | ||
| BC019276.1 | - | 5 | 558 | 2822781 | ||
| BM008944.1 | - | 5 | 514 | |||
| CF127298.1 | - | 5 | 556 | |||

NDUFB8
Genomic Map