siRNA Details
| Gene Symbol: NDUFB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005005.2 | NP_004996.1 | 1, 2 | 187 | ||
| GenBank mRNA | AF044956.1 | - | 1, 2 | 181 | ||
| AF067168.1 | - | 1, 2 | 181 | |||
| AF261090.1 | - | 1, 2 | 186 | |||
| AI907018.1 | - | 1, 2 | 204 | |||
| AK026538.1 | - | 1, 2 | 170 | |||
| AK302286.1 | - | 1, 2 | 149 | |||
| AK313432.1 | - | 1, 2 | 149 | |||
| AL136714.1 | - | 1, 2 | 157 | |||
| BC007672.2 | - | 1, 2 | 154 | 3160562 | ||
| BQ685604.1 | - | 1, 2 | 137 | |||

NDUFB9
Genomic Map