siRNA Details
| Gene Symbol: NHLH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005598.3 | NP_005589.1 | 2 | 463 | ||
| GenBank mRNA | AK313700.1 | - | 2 | 463 | ||
| BC013789.1 | - | 2 | 450 | 3866861 | ||
| BT006859.1 | - | 1 | 21 | |||
| DA612092.1 | - | 2 | 463 | |||
| M96739.1 | - | 2 | 465 | |||

NHLH1
Genomic Map