siRNA Details
| Gene Symbol: NNAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322802.1 | NP_001309731.1 | 3 | 286 | ||
| NM_005386.3 | NP_005377.1 | 3 | 291 | |||
| NM_181689.2 | NP_859017.1 | 2 | 210 | |||
| GenBank mRNA | AB002392.1 | - | 3 | 204 | ||
| AI879211.1 | - | 2 | 166 | |||
| AK312069.1 | - | 3 | 244 | |||
| BC001768.1 | - | 3 | 235 | 3353944 | ||
| BE255132.1 | - | 3 | 261 | |||
| BG472075.1 | - | 3 | 282 | |||
| BT019426.1 | - | 3 | 165 | |||
| CN388981.1 | - | 3 | 231 | |||
| HY021161.1 | - | 2 | 211 | |||
| U25033.1 | - | 3 | 234 | |||
| U25034.1 | - | 2 | 155 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001291129, NM_001291130, NM_001291128, NM_010923, NM_180960, and others.

NNAT
Genomic Map