siRNA Details
| Gene Symbol: NME3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002513.2 | NP_002504.2 | 4 | 553 | ||
| XM_011522504.2 | XP_011520806.1 | 4 | 600 | |||
| GenBank mRNA | AK308663.1 | - | 4 | 374 | ||
| BC000250.1 | - | 4 | 553 | 3357930 | ||
| BG911570.1 | - | 4 | 377 | |||
| U29656.1 | - | 4, 5 | 396 | |||

NME3
Genomic Map