siRNA Details
| Gene Symbol: NPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000905.3 | NP_000896.1 | 3 | 296 | ||
| GenBank mRNA | AI198311.1 | - | 2 | 277 | ||
| BC029497.1 | - | 3 | 296 | 5278692 | ||
| K01911.1 | - | 3 | 294 | |||
| M15789.1 | - | 2 | 222 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NPY| Gene Symbol: NPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000905.3 | NP_000896.1 | 3 | 296 | ||
| GenBank mRNA | AI198311.1 | - | 2 | 277 | ||
| BC029497.1 | - | 3 | 296 | 5278692 | ||
| K01911.1 | - | 3 | 294 | |||
| M15789.1 | - | 2 | 222 | |||