siRNA Details
| Gene Symbol: NOVA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002516.3 | NP_002507.1 | 3, 4 | 440 | ||
| XM_006723230.3 | XP_006723293.1 | 4, 5 | 492 | |||
| XM_017026838.1 | XP_016882327.1 | 3, 4 | 716 | |||
| XM_017026839.1 | XP_016882328.1 | 3, 4 | 542 | |||
| XM_017026840.1 | XP_016882329.1 | 2, 3 | 299 | |||
| GenBank mRNA | AF083898.1 | - | 3, 4 | 440 | ||
| BQ631784.1 | - | 1, 2 | 21 | |||
| U70477.1 | - | 3, 4 | 411 | |||

NOVA2
Genomic Map