siRNA Details
| Gene Symbol: PAX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002584.2 | NP_002575.1 | 8 | 2068 | ||
| NM_013945.2 | NP_039236.1 | 8 | 2062 | |||
| GenBank mRNA | BC121165.2 | - | 8 | 1544 | 40121582 | |
| BC121166.2 | - | 8 | 1544 | 40121584 | ||
| X96743.1 | - | 8 | 2069 | |||

PAX7
Genomic Map