siRNA Details
| Gene Symbol: PMAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021127.2 | NP_066950.1 | 2 | 341 | ||
| GenBank mRNA | AK311943.1 | - | 2 | 306 | ||
| BC013120.1 | - | 2 | 264 | 3858958 | ||
| BC032663.1 | - | 3 | 447 | 5529754 | ||
| BF667112.1 | - | 342 | ||||
| D90070.1 | - | 2 | 296 | |||
| KU178263.1 | - | 3 | 274 | |||
| KU178264.1 | - | 2 | 123 | |||

PMAIP1
Genomic Map