siRNA Details
| Gene Symbol: PRSS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197097.2 | NP_001184026.2 | 3 | 409 | ||
| NM_001197098.1 | NP_001184027.1 | 2 | 51 | |||
| NM_002771.3 | NP_002762.2 | 2 | 72 | |||
| NM_007343.3 | NP_031369.2 | 2 | 281 | |||
| XM_011517965.1 | XP_011516267.1 | 3 | 332 | |||
| GenBank mRNA | AB298285.1 | - | 2 | 281 | ||
| AB298286.1 | - | 2 | 51 | |||
| AY052783.1 | - | 3 | 186 | |||
| BC030238.1 | - | 2 | 71 | |||
| BC069476.1 | - | 2 | 72 | 7262223 | ||
| BC069494.1 | - | 2 | 72 | 7262235 | ||
| D45417.1 | - | 2 | 59 | |||
| X15505.1 | - | 2 | 72 | |||
| X71345.1 | - | 2 | 102 | |||
| X72781.1 | - | 2 | 102 | |||

PRSS3
Genomic Map