siRNA Details
Gene Symbol: PSMA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002792.3 | NP_002783.1 | 3 | 397 | ||
GenBank mRNA | AF022815.1 | - | 3 | 266 | ||
AF054185.1 | - | 3 | 327 | |||
AK127210.1 | - | 2 | 725 | |||
AK312025.1 | - | 3 | 292 | |||
BC001895.1 | - | 3 | 330 | |||
BC004427.1 | - | 3 | 315 | 2819923 | ||
BC014999.1 | - | 3 | 325 | |||
BG575009.1 | - | 3 | 385 | |||
BI258951.1 | - | 3 | 397 | |||
BI906714.1 | - | 3 | 283 | |||
BT007165.1 | - | 3 | 242 | |||
BU632089.1 | - | 5 | 648 | |||
GQ472224.1 | - | 3 | 315 |
