siRNA Details
| Gene Symbol: PSMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002802.2 | NP_002793.2 | 8 | 846 | ||
| XM_017021470.1 | XP_016876959.1 | 8 | 2365 | |||
| XM_017021471.1 | XP_016876960.1 | 9 | 922 | |||
| GenBank mRNA | AB451311.1 | - | 1 | 798 | ||
| AB451441.1 | - | 1 | 798 | |||
| AK124201.1 | - | 1 | 857 | |||
| AK222668.1 | - | 8 | 814 | |||
| AK299121.1 | - | 7 | 722 | |||
| AK313325.1 | - | 8 | 835 | |||
| BC000512.2 | - | 1 | 832 | 2822718 | ||
| BC016368.1 | - | 1 | 826 | 4133348 | ||
| BC018859.2 | - | 1 | 799 | |||
| BC067741.1 | - | 1 | 819 | 5264945 | ||
| BC073818.1 | - | 1 | 804 | 5456334 | ||
| BT009826.1 | - | 1 | 798 | |||
| CR457044.1 | - | 1 | 798 | |||
| L02426.1 | - | 8 | 858 | |||

PSMC1
Genomic Map