siRNA Details
| Gene Symbol: PTMS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002824.4 | NP_002815.3 | 3 | 473 | ||
| XM_011520987.1 | XP_011519289.1 | 3 | 786 | |||
| GenBank mRNA | BC017025.1 | - | 3 | 192 | 3900998 | |
| BC128229.1 | - | 4 | 140 | |||
| BM451233.1 | - | 3 | 709 | |||
| BM550529.1 | - | 550 | ||||
| M24398.1 | - | 3 | 442 | |||

PTMS
Genomic Map