siRNA Details
| Gene Symbol: RAPSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005055.4 | NP_005046.2 | 7 | 1263 | ||
| NM_032645.4 | NP_116034.2 | 5 | 1086 | |||
| XM_005253042.3 | XP_005253099.1 | 6 | 1167 | |||
| XM_005253043.3 | XP_005253100.1 | 6 | 1098 | |||
| XM_011520252.1 | XP_011518554.1 | 7 | 1306 | |||
| GenBank mRNA | AF449218.1 | - | 7 | 1263 | ||
| AK310002.1 | - | 5 | 1044 | |||
| BC004196.2 | - | 5 | 1051 | 3530551 | ||
| KU178300.1 | - | 5 | 872 | |||
| KU178301.1 | - | 7 | 1185 | |||
| KU178302.1 | - | 7 | 1049 | |||
| KU178303.1 | - | 6 | 995 | |||
| KU178304.1 | - | 6 | 926 | |||
| Z33905.1 | - | 7 | 1256 | |||

RAPSN
Genomic Map