siRNA Details
| Gene Symbol: RLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005059.3 | NP_005050.2 | 3 | 475 | ||
| NM_134441.2 | NP_604390.1 | 2 | 374 | |||
| XM_017015011.1 | XP_016870500.1 | 3 | 743 | |||
| GenBank mRNA | A06925.1 | - | 2 | 343 | ||
| A17315.1 | - | 2 | 343 | |||
| AI033912.1 | - | 1 | 453 | |||
| BC126415.1 | - | 2 | 374 | 8992131 | ||
| BC126419.1 | - | 2 | 374 | 8992135 | ||
| BG206251.1 | - | 1 | 113 | |||
| X00948.1 | - | 2 | 343 | |||

RLN2
Genomic Map