siRNA Details
| Gene Symbol: RPL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000973.4 | NP_000964.1 | 4 | 389 | ||
| NM_001317771.1 | NP_001304700.1 | 4 | 432 | |||
| NM_001317782.1 | NP_001304711.1 | 3 | 557 | |||
| NM_033301.2 | NP_150644.1 | 4 | 453 | |||
| XM_017013723.1 | XP_016869212.1 | 4 | 364 | |||
| GenBank mRNA | AK289459.1 | - | 4 | 340 | ||
| AK301073.1 | - | 4 | 333 | |||
| AK309619.1 | - | 3 | 641 | |||
| BC000047.2 | - | 2 | 161 | |||
| BC000077.2 | - | 4 | 324 | 3506015 | ||
| BC030151.2 | - | 3 | 311 | |||
| BC093064.1 | - | 4 | 363 | 30716964 | ||
| BG480561.1 | - | 3 | 512 | |||
| BQ929065.1 | - | 4 | 400 | |||
| CB128790.1 | - | 4 | 453 | |||
| CR457046.1 | - | 3 | 297 | |||
| Z28407.1 | - | 4 | 340 | |||

RPL8
Genomic Map