siRNA Details
| Gene Symbol: RRAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006270.4 | NP_006261.1 | 5 | 553 | ||
| GenBank mRNA | AF493920.1 | - | 5 | 508 | ||
| BC016286.1 | - | 5 | 532 | 3910343 | ||
| BC016318.1 | - | 5 | 557 | 4073829 | ||
| BT006805.1 | - | 5 | 508 | |||
| CR541944.1 | - | 5 | 508 | |||
| CR541967.1 | - | 5 | 508 | |||

RRAS
Genomic Map