siRNA Details
| Gene Symbol: S100A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002961.2 | NP_002952.1 | 2 | 159 | ||
| NM_019554.2 | NP_062427.1 | 3 | 208 | |||
| GenBank mRNA | AK292083.1 | - | 2 | 159 | ||
| AY762982.1 | - | 62 | ||||
| BC016300.1 | - | 2 | 163 | 4247807 | ||
| BC024974.1 | - | 2 | 143 | |||
| CB109749.1 | - | 3 | 208 | |||
| CR450345.1 | - | 1 | 90 | |||
| M80563.1 | - | 2 | 225 | |||

S100A4
Genomic Map