siRNA Details
| Gene Symbol: CXCL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002993.3 | NP_002984.1 | 4 | 830 | ||
| GenBank mRNA | BC013744.1 | - | 4 | 665 | 4471095 | |
| U81234.1 | - | 4 | 698 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CXCL6| Gene Symbol: CXCL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002993.3 | NP_002984.1 | 4 | 830 | ||
| GenBank mRNA | BC013744.1 | - | 4 | 665 | 4471095 | |
| U81234.1 | - | 4 | 698 | |||