siRNA Details
| Gene Symbol: SKP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006930.3 | NP_008861.2 | 5 | 1473 | ||
| NM_170679.2 | NP_733779.1 | 6 | 787 | |||
| GenBank mRNA | AK057110.1 | - | 2 | 1890 | ||
| BC009839.2 | - | 6 | 671 | 4053593 | ||
| BC020798.1 | - | 6 | 722 | 4243711 | ||
| BC025673.1 | - | 5 | 1370 | 5203008 | ||
| BC065730.1 | - | 6 | 708 | 6672613 | ||
| BI552966.1 | - | 6 | 755 | |||
| U37558.1 | - | 5 | 569 | |||
| Z47087.1 | - | 6 | 717 | |||

SKP1
Genomic Map