siRNA Details
| Gene Symbol: SLC8A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015063.2 | NP_055878.1 | 2 | 571 | ||
| XM_005259172.1 | XP_005259229.1 | 2 | 380 | |||
| GenBank mRNA | AB029010.1 | - | 2 | 386 | ||
| AK302552.1 | - | 2 | 571 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC8A2| Gene Symbol: SLC8A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015063.2 | NP_055878.1 | 2 | 571 | ||
| XM_005259172.1 | XP_005259229.1 | 2 | 380 | |||
| GenBank mRNA | AB029010.1 | - | 2 | 386 | ||
| AK302552.1 | - | 2 | 571 | |||