siRNA Details
| Gene Symbol: SNAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003083.3 | NP_003074.1 | 4 | 687 | ||
| NR_030717.1 | 4 | 584 | ||||
| GenBank mRNA | AK222926.1 | - | 4 | 658 | ||
| AK314878.1 | - | 4 | 672 | |||
| BC011868.2 | - | 4 | 642 | 4545178 | ||
| BX647416.1 | - | 4 | 551 | |||
| CF454397.1 | - | 4 | 687 | |||
| U44755.1 | - | 4 | 671 | |||
| U44898.1 | - | 4 | 653 | |||

SNAPC2
Genomic Map